Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IDH3A Mouse anti-Human, Canine, PE/Atto594, Clone: OTI2F11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270992PEATT594

Catalog No. N270992P594


Add to cart

Description

Description

IDH3A Monoclonal antibody specifically detects IDH3A in Human, Canine samples. It is validated for Flow Cytometry.
Specifications

Specifications

IDH3A
OTI2F11
Flow Cytometry
Affinity Purified, Conjugated
IDH3A
Full length recombinant protein of human IDH3A (NP_005521) produced in HEK293T cells.
Immunogen affinity purified
Stem Cell Markers
Primary
3419.0
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
EC 1.1.1, EC 1.1.1.41, H-IDH alpha, isocitrate dehydrogenase (NAD+) alpha chain, isocitrate dehydrogenase [NAD] subunit alpha, mitochondrial, isocitrate dehydrogenase 3 (NAD+) alpha, Isocitric dehydrogenase subunit alpha, NAD(+)-specific ICDH subunit alpha, NAD(H)-specific isocitrate dehydrogenase alpha subunit, NAD+-specific ICDH
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit