Learn More
Aviva Systems Biology Corp IDI1 Antibody - middle region (ARP54735_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP54735_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IDI1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human IDI1. Alternate names: IPP1, IPPI1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61293