Learn More
IDI2 Mouse anti-Human, Clone: 1C11, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA524506
Catalog No. PIMA524506
Add to cart
Description
Description
Peptide Sequence: MSDINLDWVD RRQLQRLEEM LIVVDENDKV IGADTKRNCH LNENIEKGLL HRAFSVVLFN TKNRILIQQR SDTKVTFPGY FTDSCSSHPL YNPAELEEKD AIGVRRAAQR RLQAELGIPG EQISPEDIVF MTIYHHKAKS DRIWGEHEIC YLLLVRKNVT LNPDPSETKS ILYLSQEELW ELLEREARGE VKVTPWLRTI AERFLYRWWP HLDDVTPFVE LHKIHRV
Catalyzes the 1,3-allylic rearrangement of the homoallylic substrate isopentenyl (IPP) to its highly electrophilic allylic isomer, dimethylallyl diphosphate (DMAPP).
Specifications
Specifications
IDI2
Monoclonal
Unconjugated
IDI2
Liquid
IDI2
IDI2 (AAH17778, 1 a.a. ∽ 227 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Protein A
RUO
Antibody
Monoclonal
Human
ELISA, Western Blot
1C11
PBS with no preservative; pH 7.4
Q9BXS1
IPPI2, IPP isomerase 2, diphosphate dimethylallyl diphosphate isomerase 2, isopentenyl pyrophosphate isomerase 2, isopentenyl-diphosphate Delta-isomerase 2
Mouse
IgG2a, kappa
100 μg
-20° C
Primary
91734