Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IDNK Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546105
Description
Peptide sequence: RVVLACSALK KTYRDILTQG KDGVALKCEE SGKEAKQAEM QLLVVHLSGS Sequence homology: Human: 100%.
The function of this protein remains unknown.
Specifications
IDNK | |
Polyclonal | |
Unconjugated | |
Idnk | |
5133401N09Rik; bA522I20.2; BCRG2; C9orf103; glucokinase-like protein; gluconate kinase; gluconokinase-like protein; hGntK; Idnk; idnK gluconokinase homolog (E. coli); IDNK, gluconokinase; idnK, gluconokinase homolog; KIAA1824; probable gluconokinase; WDR22 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
414328 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5T6J7 | |
Idnk | |
Synthetic peptide directed towards the middle region of human IDNK. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction