Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

IDO-2 Mouse anti-Human,Mouse, KHL, Clone: 13-3, MilliporeSigma™

Mouse Monoclonal Antibody

Specifications

Antigen IDO-2
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Conjugate KHL
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF23151MI
View Documents
MilliporeSigma
MABF2315100UG
100 μg
Each for $372.00
N/A
MABF23152MI
View Documents
MilliporeSigma
MABF231525UG
25 μg
Each for $147.00
N/A
Description

Description

IDO-2 Monoclonal antibody specifically detects IDO-2 in Human,Mouse samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting.
Specifications

Specifications

IDO-2
KHL
Mouse
Indoleamine 2;3-dioxygenase 2;Indoleamine 2;3-dioxygenase-like protein 1;Indoleamine-pyrrole 2;3-dioxygenase-like protein 1
KLH-conjugated peptide corresponding to 21 amino acids from the C-terminal half of human Indoleamine 2,3-dioxygenase 2 (IDO2).
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Purified
Human, Mouse
IDO2, INDOL1
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit