Learn More
Selleck Chemical LLCSupplier Diversity Partner IDO inhibitor 1 DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S85575MG
IDO inhibitor 1 is a potent inhibitor of IDO (indoleamine-(2,3)-dioxygenase) with IC50 of 3 nM.
Catalog No. 50-194-8337