Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IDO2 Mouse anti-Human, Clone: OTI1A4, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA501378

Catalog No. 50-167-2521


Add to cart

Description

Description

Along with the enzymes encoded by the INDO and TDO2 genes, the enzyme encoded by the INDOL1 gene metabolizes tryptophan in the kynurenine pathway.
Specifications

Specifications

IDO2
Monoclonal
0.5 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q6ZQW0
INDOL1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
169355
Flow Cytometry, Western Blot
OTI1A4
Unconjugated
IDO2
Liquid
IDO2
Full length human recombit protein of human IDO2 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit