Learn More
IDO2 Mouse anti-Human, Clone: OTI2D11, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF806621
Catalog No. 50-166-7927
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Along with the enzymes encoded by the INDO and TDO2 genes, the enzyme encoded by the INDOL1 gene metabolizes tryptophan in the kynurenine pathway.
Specifications
Specifications
IDO2
Monoclonal
Unconjugated
IDO2
Lyophilized
IDO2
Full length human recombit protein of human IDO2 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI2D11
PBS with 8% trehalose and no preservative; pH 7.3
Q6ZQW0
INDOL1
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
169355