Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IDO2 Mouse anti-Mouse, Alexa Fluor™ 350, Clone: 1HC, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP221641AF350

Catalog No. NP221641A30


Add to cart

Description

Description

IDO2 Monoclonal antibody specifically detects IDO2 in Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

IDO2
1HC
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Conjugated, Purified
IDO2
Anti-IDO2 (MOUSE) Monoclonal Antibody was produced in mouse by repeated immunizations with complete mouse IDO2 protein followed by hybridoma development.
Protein G purified
Store at 4°C in the dark.
Monoclonal
This antibody is specific for mouse IDO2 protein.
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
EC 1.13.11.-, IDO-2, INDOL1indoleamine-pyrrole 2,3 dioxygenase-like 1, indoleamine 2,3-dioxygenase 2, indoleamine 2,3-dioxygenase-like 1 protein, Indoleamine 2,3-dioxygenase-like protein 1, Indoleamine-pyrrole 2,3-dioxygenase-like protein 1
Mouse
IgG2a Kappa
0.1 mL
Primary
169355.0
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit