Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Iduronate 2-Sulfatase/IDS Mouse anti-Human, Mouse, Rat, Monkey, APC, Clone: OTI1D9, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270994APC
Description
Iduronate 2-Sulfatase/IDS Monoclonal specifically detects Iduronate 2-Sulfatase/IDS in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry.Specifications
Iduronate 2-Sulfatase/IDS | |
Monoclonal | |
APC | |
PBS with 0.05% Sodium Azide | |
IDS | |
Full length recombinant protein of human Iduronate 2-Sulfatase/IDS (NP_000193) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Monkey | |
IgG2a |
Flow Cytometry | |
OTI1D9 | |
Flow Cytometry | |
Alpha-L-iduronate sulfate sulfatase, EC 3.1.6.13, iduronate 2-sulfatase, iduronate 2-sulfatase 14 kDa chain, iduronate 2-sulfatase 42 kDa chain, idursulfase, MPS2, S, SIDS | |
Mouse | |
Affinity Purified | |
Proteases & Other Enzymes | |
3423.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title