Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iduronate 2-Sulfatase/IDS Mouse anti-Human, Mouse, PE/Cy5.5, Clone: OTI1D9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270994PECY55

Catalog No. N270994PEC5


Add to cart

Description

Description

Iduronate 2-Sulfatase/IDS Monoclonal antibody specifically detects Iduronate 2-Sulfatase/IDS in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

Iduronate 2-Sulfatase/IDS
OTI1D9
Flow Cytometry
Affinity Purified, Conjugated
IDS
Full length recombinant protein of human Iduronate 2-Sulfatase/IDS (NP_000193) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Mouse
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
Alpha-L-iduronate sulfate sulfatase, EC 3.1.6.13, iduronate 2-sulfatase, iduronate 2-sulfatase 14 kDa chain, iduronate 2-sulfatase 42 kDa chain, idursulfase, MPS2, S, SIDS
Mouse
IgG2a
0.1 mL
Primary
3423.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit