Learn More
Brady™ IDXPERT™ B-499 Nylon Cloth Labels
Printable, permanent indoor/outdoor grade nylon cloth label for IDXPERT printers. Brady™ IDXPERT labels are effective on smooth, rough and textured surfaces.
$84.81 - $126.49
Specifications
Product Type IDXPERT Label
Material Nylon Cloth (B-499)
Color Black, White
Format Cartridge
Adhesive Type Acrylic, Permanent
View More Specs
Products 6
Catalog Number Mfr. No. Length (English) Width (English) Tape Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Width (English) Tape Price Quantity & Availability  
19-107-909
View Documents Promotion Details
Brady™
XC1000499
20 ft. 1 in.
Each for $126.49
Add to cart
19-107-920
View Documents Promotion Details
Brady™
XC375499
20 ft. 0.375 in.
Each for $113.29
Add to cart
19-107-921
View Documents Promotion Details
Brady™
XC500499
20 ft. 0.5 in.
Each for $119.00
Add to cart
19-107-925
View Documents Promotion Details
Brady™
XC750499
20 ft. 0.75 in.
Each for $123.04
Add to cart
19-108-975
View Documents Promotion Details
Brady™
XF151499
1 in. 2.25 in.
Each for $84.81
Add to cart
19-108-976
View Documents Promotion Details
Brady™
XF152499
1.5 in. 1.5 in.
Each for $111.95
Add to cart
Description
Description
  • Printable nylon cloth labels for the IDXPERT printer
  • Material is UL recognized with its respective ribbon
Specifications
Specifications
IDXPERT Label
Black, White
Acrylic, Permanent
Compatible surface: rough, smooth and textured
iDXPERT Printer
Nylon Cloth (B-499)
Cartridge
0.0065 in.
UL Recognized, CSA Accepted, Halogen Free (DIN VDE 0472/part 815)
Cable and wire marking, Panel labeling, Voice and Data ID