Learn More
Brady™ IDXPERT™ and LABXPERT™ General Identification Labels
Printable, permanent indoor/outdoor grade vinyl tape for IDXPERT and LABXPERT printers. Brady™ General Identification Labels are made of vinyl film with permanent adhesive.
$158.60 - $162.80
Specifications
Description Compatible surface: Powder Coated, Rough, Smooth and Textured
Adhesive Type Acrylic, Permanent, Cold Temperature
Material Vinyl (B-595)
Product Type IDXPERT and LABXPERT Label
For Use With (Application) Panel Identification, Voice/Data Identification, General and Industrial Identification, Laboratory Identification
View More Specs
Products 15
Catalog Number Mfr. No. Color Size (English) Size (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Color Size (English) Size (Metric) Price Quantity & Availability  
19-107-926
View Documents
Brady™
XC750595BLWT
White, Blue 30 ft. x 0.75 in. (L x W) 9.1 m x 19.1 mm
Each for $162.80
Add to Cart
19-107-927
View Documents
Brady™
XC750595CLBK
Black, Clear 25 ft. x 0.75 in. (L x W) 6 m x 19.1 mm
Each for $158.60
Add to Cart
19-107-910
View Documents
Brady™
XC1000595BLWT
White, Blue 30 ft. x 1 in. 9.1 m x 25.4 mm Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. N/A
19-107-912
View Documents
Brady™
XC1000595YLBK
Black, Yellow 30 ft. x 1 in. 9.1 m x 25.4 mm Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. N/A
19-107-911
View Documents
Brady™
XC1000595WTRD
White, Red 30 ft. x 1 in. 9.1 m x 25.4 mm Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. N/A
19-107-916
View Documents
Brady™
XC1500595CLBK
Black, Clear 25 ft. x 1.5 in. 7.6 m x 38.1 mm Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. N/A
19-107-922
View Documents
Brady™
XC500595CLBK
Black, Clear 25 ft. x 0.5 in. (L x W) 7.6 m x 12.7 mm Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. N/A
19-107-923
View Documents
Brady™
XC500595WTBK
Black, White 30 ft. x 0.5 in. (L x W) 9.1 m x 12.7 mm Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. N/A
19-107-928
View Documents
Brady™
XC750595GNWT
White, Green 30 ft. x 0.75 in. (L x W) 9.1 m x 19.1 mm Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. N/A
19-107-929
View Documents
Brady™
XC750595ORBK
Black, Orange 30 ft. x 0.75 in. (L x W) 9.1 m x 19.1 mm Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. N/A
19-107-931
View Documents
Brady™
XC750595WTBK
Black, White 30 ft. x 1.5 in. 9.1 m x 19.1 mm Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. N/A
19-107-930
View Documents
Brady™
XC750595RDWT
White, Red 30 ft. x 1.5 in. 9.1 m x 38.1 mm Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. N/A
19-107-932
View Documents
Brady™
XC750595WTRD
White, Red 30 ft. x 0.75 in. (L x W) 9.1 m x 19.1 mm Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. N/A
19-107-933
View Documents
Brady™
XC750595YLBK
Black, Yellow 30 ft. x 0.75 in. (L x W) 9.1 m x 19.1 mm Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. N/A
19-107-915
View Documents
Brady™
XC1500595BLWT
White, Blue 30 ft. x 1.5 in. 9.1 m x 38.1 mm Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. N/A
Description
Description
  • Printable, permanent indoor/outdoor grade vinyl tape for the IDXPERT and LABXPERT printers
  • Available in a variety of colors and sizes
Specifications
Specifications
Compatible surface: Powder Coated, Rough, Smooth and Textured
Vinyl (B-595)
Panel Identification, Voice/Data Identification, General and Industrial Identification, Laboratory Identification
0.004 in.
Acrylic, Permanent, Cold Temperature
IDXPERT and LABXPERT Label
iDXPERT Printer and LabXpert Label Printer