Learn More
IEM 1754 dihydrobromide, Tocris™
Voltage-dependent, open-channel blocker of AMPA currents
Manufacturer:  R&D Systems 4199/10
Quantity 10mg
 View more versions of this product
Catalog No. 41-991-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>98%
412.25
IEM 1754 dihydrobromide
C16H30N2·2HBr
N-(Tricyclo[3.3.1.13, 7]dec-1-ylmethyl)-1, 5-pentanediamine dihydrobromide
Specifications
10mg
>98%
412.25
IEM 1754 dihydrobromide
162831-31-4
C16H30N2·2HBr
N-(Tricyclo[3.3.1.13, 7]dec-1-ylmethyl)-1, 5-pentanediamine dihydrobromide
Safety and Handling
Recommended Storage : Store at Room Temperature