Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IEM 1754 dihydrobromide, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Voltage-dependent open-channel blocker of AMPA receptors. Selective between subtypes; blocks GluA2 subunit-lacking (Ca2+-permeable) receptors more potently than GluA2-containing receptors. Also potently blocks NMDA receptor channels. Less sensitive than IEM 1460 (Cat. No. 1460) at a holding potential between -40 and -80mV.
Specifications
Quantity | 50 mg |
Synonym | iem 1754 dihydrobromide, iem1754 dihydrobromide, iem 1754 2hbr, iem1754 2 hbr, 1-amino-5-1-adamantylmethylamino-pentane dihydrobromide, n1-adamantan-1-ylmethyl pentane-1,5-diamine dihydrobromide, adamantan-1-ylmethyl 5-aminopentyl amine dihydrobromide, n-tricyclo 3.3.1.13,7 dec-1-ylmethyl-1,5-pentanediamine dihydrobromide, 5-aminopentyl 3r,5s,7s-adamantan-1-ylmethyl amine dihydrobromide |
Formula Weight | 412.25 |
Percent Purity | >98% |
Chemical Name or Material | IEM 1754 dihydrobromide |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction