Learn More
IER2 Rabbit anti-Human, Polyclonal, Bioss Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  BiossSupplier Diversity Partner BS15544R
Catalog No. 50-198-3867
Add to cart
Description
Description
DNA-binding protein that seems to act as a transcription factor. Involved in the regulation of neuronal differentiation, acts upon JNK-signaling pathway activation and plays a role in neurite outgrowth in hippocampal cells. May mediate with FIBP FGF-signaling in the establishment of laterality in the embryo. Promotes cell motility, seems to stimulate tumor metastasis.
Specifications
Specifications
IER2
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
IER2
Liquid
Rabbit
IgG
100 μL
-20°C
Primary
9592
IER2
1 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Q9BTL4
chxI protein homolog; ETR101; IER2; immediate early response 2; Immediate early response gene 2 protein; PIP92; Protein ETR101
KLH conjugated synthetic peptide derived from human IER2.
Protein A
RUO
Antibody
Polyclonal
Human