Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IER5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544655
Description
Peptide sequence: MEFKLEAHRI VSISLGKIYN SRVQRGGIKL HKNLLVSLVL RSARQVYLSD Sequence homology: Cow: 100%; Dog: 77%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 100%.
This gene encodes a protein that is similar to other immediate early response proteins. In the mouse, a similar gene may play an important role in mediating the cellular response to mitogenic signals. Studies in rats found the expression of a similar gene to be increased after waking and sleep deprivation.
Specifications
IER5 | |
Polyclonal | |
Unconjugated | |
IER5 | |
Ier5; immediate early response 5; immediate early response gene 5 protein; immediately early response; MGC102760; PP4583; SBBI48; si:dz129i22.1; Unknown (protein for MGC:136422); wu:fa99a02; wu:fb04b03; wu:fb11f08; zgc:136422 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51278 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5VY09 | |
IER5 | |
Synthetic peptide directed towards the N-terminal of human IER5 (aa 1-50). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction