Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IER5L Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP17961020UL

 View more versions of this product

Catalog No. NBP17961020


Add to cart

Description

Description

IER5L Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

IER5L
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
IER5L
Synthetic peptide directed towards the middle region of human IER5LThe immunogen for this antibody is IER5L. Peptide sequence SNLISIFGSGFSGLVSRQPDSSEQPPPLNGQLCAKQALASLGAWTRAIVA.
Immunogen affinity purified
RUO
Primary
389792
Western Blot
Western Blot 1:1000
NP_982258
bA247A12.2, immediate early response 5-like, immediate early response gene 5-like protein, MGC70833
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit