Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IEX-1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115393
Description
Antibody detects endogenous levels of total IEX1.
IEX-1 is a stress inducible gene that is induced by ionizing radiation, ultraviolet radiation, and a variety of growth factors, i.e., FAS and TNF-a. IEX-1 is widely expressed in epithelial and endocrine tissues, as well as in vascular endothelium. It plays an important role in the regulation of cellular growth, cell death and oncogenesis. IEX-1 is precisely regulated by multiple transcription factors such as p53, NF-kappa-B/rel, Sp1 and c-Myc, to ensure rapid and transient expression of IEX-1 in cells under a variety of stress conditions. IEX-1 is expressed as both a longer form (IEX1L) and a splice variant, designated IEX1S. It is localized to the nucleus and perinuclear region. Overexpression of IEX-1 facilitates apoptosis and cell cycle progression, whereas disruption of IEX-1 expression is associated with decreases in both apoptosis and cell cycle progression.
Specifications
IEX-1 | |
Polyclonal | |
Unconjugated | |
IER3 | |
AI663993; anti-death protein; cAMP inducible gene 3; cI-3; DADB-118P11.5; Dif2; DIF-2; differentiation-dependent gene 2 protein; expressed in pancreatic carcinoma; GLY96; gly96, mouse, homolog of; IER3; IEX1; IEX-1; IEX-1L; immediate early protein GLY96; immediate early response 3; immediate early response 3 protein; immediately early gene X-1; Ler3; PACAP-responsive gene 1 protein; PRG1; Protein DIF-2; Protein PRG1; Radiation-inducible immediate-early gene IEX-1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
8870 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
P46695 | |
IER3 | |
A synthesized peptide derived from human IER3(Accession P46695), corresponding to amino acid residues E112-F156. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction