Learn More
Biolog IncSupplier Diversity Partner Biolog PM M1 Inoculating Fluids DFS Item
Manufacturer:  Biolog IncSupplier Diversity Partner 72301
Similar to RPM1 1640 media but without phenol red, glucose and glutamine, 100 ml for approximately 20 microplates
Catalog No. NC0063697