Learn More
Invivogen IFA, 10 mL
Supplier: Invivogen VACIFA10
IFA (Incomplete Freund’s adjuvant) is one of the most commonly used adjuvants in research. It is prepared from non-metabolizable oils. IFA can be used to produce water-in-oil emulsions of antigens. IFA induces a predominantly Th2-biased response through the formation of a depot at the injection site and the stimulation of antibody producing plasma cells.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.