Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFFO Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | IFFO |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP152939
|
Novus Biologicals
NBP152939 |
100 μL |
Each for $436.00
|
|
NBP15293920
|
Novus Biologicals
NBP15293920UL |
20 μL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
IFFO Polyclonal specifically detects IFFO in Human samples. It is validated for Western Blot.Specifications
IFFO | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
25900 | |
Synthetic peptides corresponding to HOM-TES-103 The peptide sequence was selected from the N terminal of HOM-TES-103. Peptide sequence MGGRKRERKAAVEEDTSLSESEGPRQPDGDEEESTALSINEEMQRMLNQL. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
FLJ20703, intermediate filament family orphan 1, intermediate filament-like MGC:2625 | |
IFFO1 | |
IgG | |
Affinity Purified | |
23 kDa |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title