Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFFO1 Polyclonal Antibody
IFFO1 Polyclonal Antibody
Supplier: Thermo Scientific PA564413
Description
IFFO1 Polyclonal Antibody for IHC

Specifications
IFFO1 | |
Polyclonal | |
Unconjugated | |
IFFO1 | |
4733401N06Rik; A930037G23Rik; HOM-TES-103; Iffo; IFFO1; Intermediate filament family orphan 1; intermediate filament-like MGC:2625; RGD1308257; Tumor antigen HOM-TES-103 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25900 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q0D2I5 | |
IFFO1 | |
Recombinant protein corresponding to Human IFFO1. Recombinant protein control fragment (Product #RP-104049). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction