Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFFO2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$177.16 - $436.00

Specifications

Antigen IFFO2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094646
View Documents
Novus Biologicals
NBP305222100UL
100 μL
Each for $436.00
Add to cart
 
NB094647
View Documents
Novus Biologicals
NBP30522220UL
20 μL
Each for $177.16
Add to cart
 
Description

Description

IFFO2 Polyclonal specifically detects IFFO2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

IFFO2
Polyclonal
Purified
RUO
PBS with 50% glycerol, pH7.3.
126917
IgG
Affinity purified
Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
intermediate filament family orphan 2
Recombinant protein of human IFFO2
Primary
Store at -20°C. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit