Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFI35 Mouse anti-Human, Allophycocyanin/Cy7, Clone: OTI1C9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271004APCCY7

Catalog No. N27104APCY7


Add to cart

Description

Description

IFI35 Monoclonal antibody specifically detects IFI35 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

IFI35
OTI1C9
Flow Cytometry
Affinity Purified, Conjugated
IFI35
Full length recombinant protein of human IFI35 (NP_005524)produced in HEK293T cells
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
FLJ21753, Ifi-35, IFP 35, IFP35interferon-induced 35 kDa protein, interferon-induced protein 35
Mouse
IgG2a
0.1 mL
Primary
3430.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit