Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFI35 Mouse anti-Human, Clone: OTI1C9, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA503386

Catalog No. 50-167-4291


Add to cart

Description

Description

Not yet known.
Specifications

Specifications

IFI35
Monoclonal
0.4 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P80217
IFP35
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3430
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI1C9
Unconjugated
IFI35
Liquid
IFI35
Full length human recombit protein of human IFI35 produced in HEk293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit