Learn More
IFIH1 Mouse anti-Human, Clone: OTI3E10, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF803668
Catalog No. 50-166-6619
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Melanoma differentiation associated gene 5, MDA5, is a pattern recognition receptor to transmit the signal.Once activated, both proteins signal through IPS-1 activating transcription factors NF- kappaB and IRF-3 and ultimately activating apoptosis, cytokine signaling, and inflammation. RIG-I is essential for signaling by influenza A, influenza B, human respiratory syncytial virus, paromyxoviruses, Japanese encephalitis virus. MDA5 is essential for signaling by picornavirus. Both RIG-I and MDA5 are responsible for signaling by West Nile virus.
Specifications
Specifications
IFIH1
Monoclonal
Unconjugated
IFIH1
Lyophilized
IFIH1
Full length human recombit protein of human IFIH1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI3E10
PBS with 8% trehalose and no preservative; pH 7.3
Q9BYX4
AGS7, Hlcd, IDDM19, MDA-5, MDA5, RLR-2, SGMRT1
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
64135