Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFIT1 Mouse anti-Human, Mouse, Rat, Monkey, PE/Cy5.5, Clone: OTI3G8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271005PECY55

Catalog No. N27105PECY5


Add to cart

Description

Description

IFIT1 Monoclonal antibody specifically detects IFIT1 in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

IFIT1
OTI3G8
Flow Cytometry
Affinity Purified, Conjugated
IFIT1
Full length recombinant protein of human IFIT1 (NP_000899) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Monkey, Mouse, Rat
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
G10P1C56, GARG-16, IFI56IFI-56, IFI-56K, IFIT-1, IFNAI1ISG56, Interferon, alpha-inducible protein (MW 56kD), Interferon-induced 56 kDa protein, interferon-induced protein 56, interferon-induced protein with tetratricopeptide repeats 1, p56, RNM561
Mouse
IgG1
0.1 mL
Primary
3434.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit