Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFIT3 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 152011AP

Catalog No. 50-173-0061


Add to cart

Description

Description

The function remains unknown.
Specifications

Specifications

IFIT3
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O14879
CIG 49, CIG49, GARG 49, IFI 60K, IFI60, IFIT 3, IFIT 4, IFIT3, IFIT4, IRG2, ISG 60, ISG60, RIG G
Rabbit
IgG
150 μL
-20°C
Primary
3437
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
IFIT3
Liquid
IFIT3
IFIT3 Fusion Protein Ag7349
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit