Learn More
Abnova™ IFITM1 293T Cell Transient Overexpression Lysate (Denatured) (T04)
Human overexpression lysate
Manufacturer:  Abnova™ H00008519T04
Catalog No. 89-948-306
Add to cart
Specifications
Specifications
Western Blot
293T
BC000897
pCMV-IFITM1 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
IFITM1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 13.9kDa
Transient overexpression cell lysate was tested with anti-IFITM1 antibody (Mfr. No. H00008519-B02) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)