Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFITM2 Monoclonal Antibody (OTI4E3), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA527504
Description
IFITM2 Monoclonal antibody specifically detects IFITM2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
IFITM2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q01629 | |
IFITM2 | |
Full length human recombinant protein of IFITM2 produced in E.coli | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4E3 | |
Unconjugated | |
IFITM2 | |
1-8D; dispanin subfamily A member 2c; DSPA2c; IFITM2; ifm2; interferon induced transmembrane protein 2; interferon-induced transmembrane protein 2; Interferon-inducible protein 1-8D | |
Mouse | |
Affinity Chromatography | |
RUO | |
10581 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction