Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFLTD1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551311

Catalog No. PA551311


Add to cart

Description

Description

The antibody detects endogenous levels of total IFLTD1 / LMNTD1 protein.

The specific function of this protein remains unknown.
Specifications

Specifications

IFLTD1
Polyclonal
Unconjugated
LMNTD1
Liquid
LMNTD1
Fusion protein of human LMNTD1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
0.8 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q8N9Z9
IFLTD1
Rabbit
IgG
100 μL
-20°C
Primary
160492
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit