Learn More
Aat BioquestSupplier Diversity Partner iFluor™ 488 succinimidyl ester DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 1023
Although FITC is still the most popular fluorescent labeling dye for preparing green fluorescent bioconjugates, there are certain limitations with FITC, such as severe photobleaching for microscope imaging and pH-sensitive fluorescence.
Catalog No. 50-195-2964