Learn More
Aat BioquestSupplier Diversity Partner iFluor™ 594 succinimidyl ester DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 1029
AAT Bioquest's iFluor™ dyes are optimized for labeling proteins, in particular, antibodies. iFluor™ 594 dyes have fluorescence excitation and emission maxima of ~590 nm and ~610 nm respectively.
Catalog No. 50-195-2968