Learn More
Aat BioquestSupplier Diversity Partner iFluor™ 647 alkyne DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 1090
AAT Bioquest's iFluor™ dyes are optimized for labeling proteins, in particular, antibodies. These dyes are bright, photostable and have minimal quenching on proteins.
Catalog No. 50-195-3002