Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-alpha 1 Mouse anti-Human, Alexa Fluor™ 594, Clone: 144BS, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP229374AF594

Catalog No. NP229374A94


Add to cart

Description

Description

IFN-alpha 1 Monoclonal antibody specifically detects IFN-alpha 1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen).
Specifications

Specifications

IFN-alpha 1
144BS
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Affinity Purified, Conjugated
Mouse
IgG1 Kappa
0.1 mL
Primary
3439.0
Human
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
IFNA1
This antibody was raised against Interferon alpha.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
This antibody binds but does not neutralize the activity of human IFN-alpha subtypes - B, -C, -D and -F, as well as several IFN-alpha B/D, D/B hybrids.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit