Learn More
IFN-alpha 1 Mouse anti-Human, Alexa Fluor™ 750, Clone: 144BS, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP229374AF750
Catalog No. NP229374A70
Add to cart
Description
Description
IFN-alpha 1 Monoclonal antibody specifically detects IFN-alpha 1 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Frozen).
Specifications
Specifications
IFN-alpha 1
144BS
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
IFNA1
This antibody was raised against Interferon alpha.
0.1 mL
Primary
3439
Human
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
This antibody binds but does not neutralize the activity of human IFN-alpha subtypes - B, -C, -D and -F, as well as several IFN-alpha B/D, D/B hybrids.