Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-alpha 2 Antibody (ST29), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002479AF405
Description
IFN-alpha 2 Monoclonal antibody specifically detects IFN-alpha 2 in Human samples. It is validated for ELISASpecifications
IFN-alpha 2 | |
Monoclonal | |
Alexa Fluor 405 | |
alpha-2a interferon, IFNA, IFN-alpha-2, IFN-alphaA, INFA2, interferon alpha 2b, interferon alpha A, interferon alpha-2, Interferon alpha-A, interferon, alpha 2, LeIF A, MGC125764, MGC125765 | |
Human interferon (HuIFN) produced by Namalwa cells (HuIFN-aN) | |
0.1 mL | |
Apoptosis, Biologically Active Proteins, Cytokine Research | |
3440 | |
Store at 4°C in the dark. | |
IgG1 |
ELISA | |
ST29 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction