Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-alpha/beta R1 Antibody (2951C), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB2452C
Description
IFN-alpha/beta R1 Monoclonal antibody specifically detects IFN-alpha/beta R1 in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
IFN-alpha/beta R1 | |
Monoclonal | |
PerCP | |
PBS | |
Rabbit | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry | |
2951C | |
Western Blot, Flow Cytometry | |
alpha-type antiviral protein, AVP, beta-type antiviral protein, CRF2-1, Cytokine receptor class-II member 1, Cytokine receptor family 2 member 1, human interferon-alpha receptor (HuIFN-alpha-Rec)10IFRC, IFN-alpha/beta receptor 1, IFN-alpha-REC, IFNAR, IFNBR, IFN-R-1, interferon (alpha, beta and omega) receptor 1, interferon alpha/beta receptor 1, interferon-alpha/beta receptor alpha chain, interferon-beta receptor 1, Type I interferon receptor 1 | |
Mouse myeloma cell line, NS0-derived human IFN-alpha/beta R1, Gly26-Lys436, Accession # P17181 | |
0.1 mL | |
Neuroscience, Phospho Specific | |
3454 | |
Store at 4C in the dark. | |
IgG |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction