Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ IFN-alpha/beta R1 Antibody Pair [HRP]
Polyclonal/Monoclonal Antibody Pair
$1183.00
Specifications
Assay Range | 125 pg/mL |
---|---|
Assay Sensitivity | 125-8000 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Immunoassay Kit Format | Sandwich ELISA Kit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB010217
|
Novus Biologicals™
NBP27958915PLATES |
15 Plates |
Each of 1 for $1,183.00
|
|
|||||
Description
This solid phase sandwich ELISA utilizes an antibody specific for Mouse IFN-alpha/beta R1.Specifications
125 pg/mL | |
Sandwich ELISA | |
Sandwich ELISA Kit | |
This solid phase sandwich ELISA utilizes an antibody specific for Mouse IFN-alpha/beta R1. | |
Sandwich ELISA | |
3454 | |
Rabbit | |
RUO | |
Storage is content dependent. | |
1. Capture Antibody: Dilute to a working concentration of 1 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 8 ng/mL is recommended. |
125-8000 pg/mL | |
HRP | |
Antibody Pairs | |
Mouse | |
alpha-type antiviral protein, AVP, beta-type antiviral protein, CRF2-1, Cytokine receptor class-II member 1, Cytokine receptor family 2 member 1, human interferon-alpha receptor (HuIFN-alpha-Rec)10IFRC, IFN-alpha/beta receptor 1, IFN-alpha-REC, IFNAR, IFNBR, IFN-R-1, interferon (alpha, beta and omega) receptor 1, interferon alpha/beta receptor 1, interferon-alpha/beta receptor alpha chain, interferon-beta receptor 1, Type I interferon receptor 1 | |
IFNAR1 | |
Neuroscience, Phospho Specific | |
IFN-alpha/beta R1 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title