Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ IFN-alpha G/IFNA5 Antibody Pair [HRP]

Polyclonal/Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2795785PLATES

 View more versions of this product

Catalog No. NB010196


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes an antibody specific for Human IFN-alpha G/IFNA5.
Specifications

Specifications

Sandwich ELISA
Antibody Pairs
39.06 pg/mL
3442
RUO
IFN-alpha G/IFNA5
5 Plates
39.06-2500 pg/mL
IFN-alpha-5, IFN-alphaG, INFA5, interferon alpha-5, Interferon alpha-61, Interferon alpha-G, interferon, alpha 5, LeIF G
Conjugated
HRP
This solid phase sandwich ELISA utilizes an antibody specific for Human IFN-alpha G/IFNA5.
Sandwich ELISA
  • Standard: Each vial contains 90 ng of recombinant Human IFN-alpha G/IFNA5.
  • Capture Antibody: 1 mg/mL of rabbit anti-Human IFN-alpha G/IFNA5 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human IFN-alpha G/IFNA5 polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Storage is content dependent.
    Rabbit
    Human
    IFNA5
    1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 2500 pg/mL is recommended.
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit