Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-alpha G/IFNA5 Rabbit anti-Human, Clone: 126, Novus Biologicals™
Rabbit Monoclonal Antibody
$321.00 - $493.00
Specifications
Antigen | IFN-alpha G/IFNA5 |
---|---|
Clone | 126 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
IFN-alpha G/IFNA5 Monoclonal specifically detects IFN-alpha G/IFNA5 in Human samples. It is validated for ELISA.Specifications
IFN-alpha G/IFNA5 | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
3442 | |
IgG | |
Protein A purified |
126 | |
ELISA | |
Unconjugated | |
Rabbit | |
Human | |
IFN-alpha-5, IFN-alphaG, INFA5, interferon alpha-5, Interferon alpha-61, Interferon alpha-G, interferon, alpha 5, LeIF G | |
Recombinant Human IFN-alpha G/IFNA5 Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title