Learn More
IFN-alpha Goat anti-Cotton Rat, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
Manufacturer:  R&D Systems AF1082SP
Catalog No. AF1082SP
Add to cart
This item is not returnable. View return policy
Description
Description
IFN-alpha Polyclonal specifically detects IFN-alpha in Cotton Rat samples. It is validated for Western Blot, Neutralization.
Specifications
Specifications
IFN-alpha
LYOPH
Western Blot 0.1 ug/mL, Neutralization 0.015-0.065 ug/mL
AAL18816
IFNA, IFNA2, IFNA2b, IFNalpha, IFN-alphaA, INFA2, interferon alpha 2, LeIF D
E. coli-derived recombinant cotton rat IFN-alpha Cys24-Lys190 (Cys109Tyr) Accession # AAL18816
Immunogen affinity purified
RUO
Primary
3440
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot, Neutralization
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Goat
IgG
25 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects cotton rat IFN-alpha in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant human IFN-beta and recombinant cotton rat IFN-gamma is observed.
Cotton Rat