Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-alpha 2 Mouse anti-Human, Allophycocyanin/Cy7, Clone: N27, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254409APCCY7

Catalog No. N254409APC7


Add to cart

Description

Description

IFN-alpha 2 Monoclonal antibody specifically detects IFN-alpha 2 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

IFN-alpha 2
N27
Flow Cytometry
Conjugated, Purified
IFNA2
Purified recombinant human IFN-alpha2
Protein A or G purified
Biologically Active Proteins
Primary
3440.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
alpha-2a interferon, IFNA, IFN-alpha-2, IFN-alphaA, INFA2, interferon alpha 2b, interferon alpha A, interferon alpha-2, Interferon alpha-A, interferon, alpha 2, LeIF A, MGC125764, MGC125765
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit