Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-alpha Mouse anti-Human, Clone: N27- Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
Antigen | IFN-alpha |
---|---|
Clone | N27 |
Concentration | 1 mg/mL |
Dilution | Flow Cytometry 0.5 - 1 ug/million cells, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, CyTOF-ready |
Applications | Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB254409200
|
Novus Biologicals
NBP254409200UG |
200 μg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
IFN-alpha Monoclonal antibody specifically detects IFN-alpha in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, CyTOF-ready.Specifications
IFN-alpha | |
1 mg/mL | |
Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF | |
Unconjugated | |
Mouse | |
Biologically Active Proteins | |
PBS with No Preservative | |
IFNA2 | |
IgG1 κ | |
Protein A or G purified | |
22 kDa |
N27 | |
Flow Cytometry 0.5 - 1 ug/million cells, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
3440 | |
Purified recombinant human IFN-alpha2 | |
Primary | |
Store at -20 to -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title