Learn More
Invitrogen™ IFN beta Human ELISA Kit
Measure levels of human interferon beta (IFNb) in human cell and tissue culture media supernatant samples using 96-well plates.
$826.00
Specifications
Description | Human IFN beta ELISA Kit |
---|---|
Immunoassay Kit Format | Colorimetric ELISA (TMB substrate) with Plates |
Specificity | No significant cross-reaction with human IFN-a or IFN-g; mouse or rat IFN-ß |
Measurement | Absorbance at 450nm |
Precision | Intra-assay CV ≤ 10%; inter-assay CV ≤ 10% |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PI414101
|
Invitrogen™
414101 |
96 Tests |
Each of 1 for $826.00
|
|
|||||
Description
Assay IFN-b (human interferon beta) levels in human cell and tissue culture media (supernatants) with theseInvitrgen™ Human IFN beta ELISA Kits.
These human IFN beta ELISA kits use pre-titered, matched pairs of coating and detection antibodies to accurately measure interferon beta (IFN-β) in samples at 100μL per well. These IFN-β kits are validated for use with human cell and tissue culture media. Also included are calibrated recombinant protein standard, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.Target: Human interferon beta (human IFN-β, IFN-beta)
Format: Colorimetric ELISA (TMB substrate) in clear 96-well plate
Measurement: Absorbance at 450nm
Assay Range: 25 to 2000pg/mL
Sensitivity: Undetermined
Precision: Intra-assay CV ≤ 10%; inter-assay CV ≤ 10%
Sample Types: Cell culture supernatant, tissue culture media
Total Assay Time: 3.25 hours
Sample Size: 100μL
Specificity: No significant cross-reaction with human IFN-α or IFN-γ; mouse or rat IFN-β
Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, substrates, and stop solution.
Specifications
Human IFN beta ELISA Kit | |
No significant cross-reaction with human IFN-a or IFN-g; mouse or rat IFN-ß | |
Intra-assay CV ≤ 10%; inter-assay CV ≤ 10% |
Colorimetric ELISA (TMB substrate) with Plates | |
Absorbance at 450nm |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.