Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-beta Rabbit anti-Human, Clone: 40, Novus Biologicals™
Rabbit Monoclonal Antibody
$321.00 - $493.00
Specifications
Antigen | IFN-beta |
---|---|
Clone | 40 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
IFN-beta Monoclonal specifically detects IFN-beta in Human samples. It is validated for ELISA.Specifications
IFN-beta | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Fibroblast interferon, IFBIFNB, IFF, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956 | |
Recombinant Human IFN-beta protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
40 | |
ELISA | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Cytokine Research, Immunology, Innate Immunity | |
0.2 um filtered solution in PBS | |
3456 | |
IgG | |
Protein A purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title