Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-gamma R2 Hamster anti-Mouse, Alexa Fluor™ 350, Clone: RM0252-7A25, Novus Biologicals™

Hamster Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212430AF350

Catalog No. NP212430A30


Add to cart

Description

Description

IFN-gamma R2 Monoclonal antibody specifically detects IFN-gamma R2 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

IFN-gamma R2
RM0252-7A25
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
IFNGR2
Mouse recombinant protein of IFNGR2, extracellular domain
Protein G purified
Store at 4°C in the dark.
Monoclonal
Mouse
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
AF-1interferon-gamma receptor beta chain precursor10IFNGT1Interferon gamma receptor accessory factor 1, IFGR2, IFN-gamma receptor 2, IFN-gamma-R2, interferon gamma receptor 2, interferon gamma receptor 2 (interferon gamma transducer 1), interferon gamma receptor accessory factor-1, interferon gamma receptor beta chain, Interferon gamma transducer 1
Hamster
IgG
0.1 mL
Primary
3460.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit