Learn More
Novus Biologicals™ IFN-gamma ELISA Kit [Biotin]
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP192669
Catalog No. NBP192669
Add to cart
Description
Description
The Mouse IFN-gamma ELISA Kit (Colorimetric) measures mouse IFN-gamma in cell culture supernatant, serum.
Specifications
Specifications
15.6 - 1,000pg/mL
3458
ELISA Kit
5.3pg/mL
Storage of components varies. See protocol for specific instructions.
IFNG
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IFN-gamma, Biotin-Conjugate anti-mouse IFN-gamma monoclonal antibody, Streptavidin-HRP, Mouse IFN-gamma Standard lyophilized, 2ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum
IFN-gamma
IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma