Learn More
IFN-gamma Mouse anti-Human, Alexa Fluor™ 350, Clone: 43-11, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP231044AF350
Catalog No. NP231044A30
Add to cart
Description
Description
IFN-gamma Monoclonal antibody specifically detects IFN-gamma in Human samples. It is validated for Western Blot,ELISA.
Specifications
Specifications
IFN-gamma
43-11
Western Blot, ELISA
IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
This antibody recognizes both the recombinant and the natural form of human interferon-gamma (IFN-gamma) (1). This antibody does not react with human IL-4, IFN-alpha or acid inactivated IFN-gamma, and it shows only a minimal reactivity with heatinactivated IFN-g (<0.05%) (1).
ELISA, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
IFNG
Monoclonal Antibody to Human IFN-g (ELISA Capture)-Azide Free
0.1 mL
Primary
3458
Human